EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Controlled synthesis of homopolymers and block copolymers based on 2-(acetoacetoxy)ethyl methacrylate via RAFT radical polymerisation.
PMID 12638988
Abstract
Reversible addition-fragmentation chain transfer (RAFT) radical polymerisation enables to synthesise well-defined homopolymers and block copolymers based on 2-(acetoacetoxy)ethyl methacrylate, which are capable to strongly coordinate to metals and metal ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17455
tert-Butyl α-bromoisobutyrate, ≥98.0%
C8H15BrO2