EMAIL THIS PAGE TO A FRIEND
Dermatology online journal
Pimozide: use in dermatology.
PMID 12639456
Abstract
Pimozide is widely used in psychiatry for chronic psychoses, schizophrenia, the syndrome of Gilles de la Tourette and to a certain extent, also in dermatology. The only dermatological indication is for delusions of parasitosis. Though there is a good rationale for using pimozide in this disease, the majority of the studies on pimozide in dermatology are uncontrolled trials and case reports.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1793 Pimozide
C28H29F2N3O