EMAIL THIS PAGE TO A FRIEND
Developmental neuroscience
Effects of selective nitric oxide synthase inhibition on IGF-1, caspases and cytokines in a newborn piglet model of perinatal hypoxia-ischaemia.
PMID 12640178
Abstract
Selective inhibition of neuronal and inducible nitric oxide synthase (NOS) with 2-iminobiotin previously showed a reduction in brain cell injury. In the present study, we investigated the effects of 2-iminobiotin treatment on insulin-like growth factor-1 (IGF-1) expression, caspase activity and cytokine expression in a newborn piglet model of perinatal hypoxia-ischaemia. Newborn piglets were subjected to 1 h of hypoxia-ischaemia and were treated intravenously with vehicle or 2-iminobiotin. Vehicle-treated piglets showed reduced IGF-1 mRNA expression and increased caspase-3 activity and DNA fragmentation. 2-Iminobiotin treatment, administered immediately upon reperfusion, prevented these observations. No differences in caspase-8 and -9 activity and cytokine [interleukin (IL)-1alpha/beta, IL-6, tumour necrosis factor (TNF)-alpha, transforming growth factor (TGF)-beta] mRNA expression were demonstrated between vehicle- and 2-iminobiotin-treated piglets at 24 h following hypoxia-ischaemia. IGF-1 mRNA correlated inversely with caspase-3 and transferase-mediated dUTP-biotin in situ nick end labelling score in the cortex, but positively with caspase-8. Cytokine mRNA did not correlate with IGF-1 mRNA, caspase-3 activity or DNA fragmentation. The present results indicate that the previously demonstrated neuroprotective effect of 2-iminobiotin treatment after perinatal hypoxia-ischaemia coincided with a preservation of the endogenous IGF-1 production and reduced caspase-3 activity, but not with a significant decrease in cytokine production.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I4632
2-Iminobiotin, ≥98% (TLC)
C10H17N3O2S