EMAIL THIS PAGE TO A FRIEND
Arzneimittel-Forschung
Suitability of cefotiam and cefuroxime axetil for the perioperative short-term prophylaxis in tonsillectomy patients.
PMID 12642969
Abstract
The efficacy of the perioperative short-term prophylaxis with cefotiam (CAS 66309-69-1) and cefuroxime axetil (CAS 64544-07-6) was analysed by the assessment of the pharmacological kinetics in the serum and the tonsil tissue in 50 patients with recurrent tonsillitis. Twenty-four patients received 1 g cefotiam by the intravenous route 30 min to 4 h before the tonsillectomy, and 26 patients received 250 mg cefuroxime axetil orally 1 to 6 h before the tonsillectomy. Bactericidal serum levels were reached for cefotiam up to 4 h after intravenous application and for cefuroxime axetil up to 3 h after oral application. In the tissue of the tonsil there were proved levels which were definitely above the MIC 90 (MIC = minimum inhibitory concentration) known for the clinically relevant germs for cefotiam after 30 min up to 2 h, for cefuroxime axetil after only 2 h. Considering the distribution areas, the capacity of the protein binding and the microbiological measuring methods, one can expect an efficient antibiotic coverage after an intravenous one-shot bolus injection of 1 g cefotiam from 30 min to 4 h and after oral application of 250 mg cefuroxime axetil on an empty stomach from 1 to 6 h. Because of the short duration of a tonsillectomy and the serum and tonsil tissue kinetics cefotiam and cefuroxime axetil are suitable for the perioperative antibiotic prophylaxis of high-risk patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0056 Cefotiam dihydrochloride hydrate, ≥98% (HPLC)
C18H23N9O4S3 · 2HCl · xH2O