EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
N-Phenyl-N-purin-6-yl ureas: the design and synthesis of p38alpha MAP kinase inhibitors.
PMID 12643941
Abstract
The design, synthesis and SAR of a series of 2,6,9-trisubstituted purine inhibitors of p38alpha kinase is reported. Synthetic routes were devised to allow for array synthesis in which all three points of diversity could be facilely explored. The binding of this novel series to p38alpha kinase, which was predicted to have several key interactions in common with SB-203580, was confirmed by X-ray crystallography of 19 (p38 IC(50)=82 nM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196614
2,6-Difluoroaniline, ≥97%
C6H5F2N