EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Room-temperature Stille cross-couplings of alkenyltin reagents and functionalized alkyl bromides that possess beta hydrogens.
PMID 12656600
Abstract
This communication describes a significant expansion in the scope of Stille reactions of Csp3-X electrophiles, specifically, that Pd/P(t-Bu)2Me catalyzes the room-temperature cross-coupling of a variety of functionalized, beta-hydrogen-containing alkyl bromides with an array of alkenyltin reagents. The structure of the phosphine (P(t-Bu)2Me) is well suited for facilitating oxidative addition and avoiding beta-hydride elimination, while the fluoride serves to activate the tin reagent for efficient transmetalation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
222380
Allylpalladium(II) chloride dimer, 98%
C6H10Cl2Pd2
95085
Tributyl(vinyl)stannane, purum, ≥97.0% (GC)
C14H30Sn