EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Neurodegeneration and neuroprotection in glaucoma: development of a therapeutic neuroprotective vaccine: the Friedenwald lecture.
PMID 12657572