EMAIL THIS PAGE TO A FRIEND
Neuroreport
Effect of steroid injection to newborn rats on serotonin activity in frontal cortex and raphe.
PMID 12657893
Abstract
The effects of injecting testosterone propionate or estradiol benzoate to newborn rats on dopaminergic and serotoninergic activity in the frontal cortex, dorsal and median raphe nucleus were analyzed when animals reached adulthood. High performance liquid chromatography was used to measure tissue levels of dopamine, serotonin and its metabolites. Activity was calculated as the metabolite/neurotransmitter ratio. An increase in androgen or estrogen levels at birth caused a significant decrease in serotoninergic activity in the frontal cortex and in the dorsal raphe nucleus, without causing apparent changes in dopaminergic activity; serotinergic activity in the median raphe nucleus was not affected. The results suggest that the transmission of DA and 5-HT in these structures are differentially influenced by early androgenization or estrogenization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5508
DL-Indole-3-lactic acid, 99%
C11H11NO3