EMAIL THIS PAGE TO A FRIEND
Organic letters
Flexibility in the partial reduction of 2,5-disubstituted pyrroles: application to the synthesis of DMDP.
PMID 12659558
Abstract
[reaction: see text] The partial reduction of electron-deficient 2,5-disubstituted pyrroles has been developed into a flexible procedure that gives control of relative stereochemistry by variation of the reduction conditions. After the reaction, the pyrroline products were dihydroxylated at C-3,4 to give either the cis or trans isomers; this flexibility means that a variety of polyhydroxylated pyrrolidines can be prepared in a short sequence. Finally, this method was applied to a synthesis of the naturally occurring glycosidase inhibitor DMDP.