EMAIL THIS PAGE TO A FRIEND
The journal of peptide research : official journal of the American Peptide Society
Simple and stereospecific homologation of urethane-protected alpha-amino acids to their higher homologs using HBTU.
PMID 12662356
Abstract
A simple, efficient and stereospecific approach for the homologation of urethane-protected alpha-amino acids to beta-amino acids by the Arndt-Eistert method employing Fmoc-/Boc-alpha-amino acid and 2-(1H-benzotriazole-1-yl)-1,1,3,3-tetramethyl-uronium hexafluorophosphate mixture for the acylation of diazomethane synthesizing the key intermediates Fmoc-/Boc-alpha-aminodiazomethanes as crystalline solids is described.