EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
Laccase-catalysed synthesis of coupling products of phenolic substrates in different reactors.
PMID 12664150
Abstract
Substrate oxidation of aromatic substances by the enzyme laccase followed by a heteromolecular coupling with a co-substrate is a promising possibility for the synthesis of new compounds. To find a suitable reactor for the effective production of new compounds, the laccase-catalysed coupling of 3-(3,4-dihydroxyphenyl)propionic acid with 4-aminobenzoic acid was investigated as a model system. Based on the kinetic parameters, a mathematical model was used to predict the reaction yield and oxygen demand in a discontinuously stirred tank reactor and a continuously operated stirred tank reactor. Membrane processes were used for bubble-free aeration of the system and to recover the soluble enzyme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
102601
3,4-Dihydroxyhydrocinnamic acid, 98%
C9H10O4