EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
PGG2, 11R-HPETE and 15R/S-HPETE are formed from different conformers of arachidonic acid in the prostaglandin endoperoxide H synthase-1 cyclooxygenase site.
PMID 12664566
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7742
Prostaglandin G2, ≥95% (HPLC), acetone solution
C20H32O6