EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Novel nonimidazole histamine H3 receptor antagonists: 1-(4-(phenoxymethyl)benzyl)piperidines and related compounds.
PMID 12672253
Abstract
In an extension of very recently published studies on successful imidazole replacements in some series of histamine H(3) receptor antagonists, we report on a new class of lipophilic nonimidazole antagonist having an aliphatic tertiary amino moiety connected to a benzyl template substituted in the 4-position by a phenoxymethyl group. The structural modifications were performed with the intention to avoid possible negative side effects reported for other series of antagonists. The novel compounds combine different characteristics of recently developed histamine H(3) receptor antagonists. The compounds were screened for their affinity in a binding assay for the human histamine H(3) receptor stably expressed in CHO-K1 cells and tested for their in vivo potency in the central nervous system of mice after oral administration. Different substitution patterns on the phenoxy group were used to optimize in vitro and/or in vivo potency leading to some compounds with low nanomolar affinity and high oral in vivo potency. Modifications of the basic piperidino moiety were performed by ring expansion, contraction, and opening. Selected compounds exhibited selectivity in functional assays on isolated organs of guinea-pig for H(3) vs H(1) and H(2) receptors. Unexpectedly, some of the novel antagonists also showed a slight preference for the human histamine H(3) receptor compared to their affinities for the guinea-pig H(3) receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7780
3-Methylhistamine dihydrochloride, analytical standard
C6H11N3 · 2HCl