EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Structural biology. A glimpse into tmRNA-mediated ribosome rescue.
PMID 12677051
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K1507
Kirromycin from Streptomyces collinus
C43H60N2O12