EMAIL THIS PAGE TO A FRIEND
Journal of molecular endocrinology
Thyrotropin-releasing hormone receptors -- similarities and differences.
PMID 12683933
Abstract
Thyrotropin-releasing hormone (TRH) initiates its effects by interacting with cell-surface membrane receptors. Two G protein-coupled receptors for TRH, TRH receptor type 1 (TRH-R1) and TRH receptor type 2 (TRH-R2), have been cloned from mammals. In this review, we compare TRH-R1 and TRH-R2 with regard to their tIssue distribution, binding affinities for TRH and TRH analogs, basal and activated signaling activities and characteristics of internalization. TRH-R1 and TRH-R2 are distributed differently in the brain and peripheral tIssues, but exhibit indistinguishable binding affinities for TRH and TRH analogs. Although they both can be stimulated by TRH to similar maximal signaling levels, TRH-R2 exhibits higher basal signaling activity and is more rapidly internalized than TRH-R1. These differences in signaling and internalization properties are probably important in the distinct parts that TRH-R1 and TRH-R2 may play in mammalian physiology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV13104
Anti-TRHR antibody produced in rabbit, IgG fraction of antiserum