EMAIL THIS PAGE TO A FRIEND
Journal of microbiological methods
A simplified dehydrogenase enzyme assay in contaminated sediment using 2-(p-Iodophenyl)-3(p-nitrophenyl)-5-phenyl tetrazolium chloride.
PMID 12689719
Abstract
2-(p-Iodophenyl)-3(p-nitrophenyl)-5-phenyl tetrazolium chloride (INT) accepts electrons from dehydrogenase enzymes and is reduced to a red-colored formazan (INTF), which can be quantified by colorimetric analysis. Use of previously published methods for this technique was unsuccessful due to background chemical reactions from high levels of polycyclic aromatic hydrocarbons (PAHs) and metals in the sediments. A modified method using acetonitrile extraction of the INTF was efficient and did not chemically reduce INT. This activity method is simple, quick, inexpensive and precise.