EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
An array of details on G-protein coupled receptor signaling: differential effects of alpha1-adrenergic receptor subtypes on gene expression and cytokine receptor signaling.
PMID 12695523