EMAIL THIS PAGE TO A FRIEND
Implant dentistry
The use of lipiodol in spiral tomography for dental implant imaging.
PMID 12704954
Abstract
Preoperative radiographic assessment is mandatory for the meticulous treatment planning and successful placement of implants. The precise topographical evaluation of selected implant sites, apart from the most straightforward cases, requires tomographic examination with the use of diagnostic radiographic templates. The incorporation of various radiopaque materials such as barium sulphate and lead foil as radiographic markers had not been entirely satisfactory. A new contrast medium, Lipiodol ethiodized oil (Laboratoire Guerbet, Paris, France), can easily be mixed with the monomer of autopolymerizing acrylic resin. The resultant acrylic template has several advantages. The tomographic images (Scanora; Soredex Orion Corporation, Helsinki, Finland) are radiographically homogenous and show the contour of the future prosthesis, the angulation of the planned implant, and the thickness of the soft tissue. In addition, the radiopaque template is optically transparent (with a slight yellow tint), which facilitates good visibility of surgical sites when the template is modified to become the surgical guide for implant placement. This transparent template can be further modified for implant position registration at the time of surgery. Because this multipurpose template is simple to construct and inexpensive, it can be recommended for routine use in implant dentistry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF96512597
Lead, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF74259793
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF57430638
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Pb
GF63249419
Lead, foil, 150x150mm, thickness 2.0mm, as rolled, 99.95%
Pb
GF60047760
Lead, foil, 250x250mm, thickness 3.0mm, as rolled, 99.95%
Pb
GF05521160
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF42741637
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Pb
GF60380940
Lead, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF85562994
Lead, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF74435519
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF10987757
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Pb
GF03985724
Lead, foil, 25x25mm, thickness 0.55mm, as rolled, 99.999%
Pb
GF73081852
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF68898188
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Pb
GF90025129
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF55764041
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.95%
Pb
GF85349346
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Pb
GF70374810
Lead, foil, 25x25mm, thickness 1.5mm, as rolled, 99.99+%
Pb
GF22900766
Lead, foil, 25x25mm, thickness 3.0mm, as rolled, 99.99+%
Pb
GF36533505
Lead, foil, 300x300mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF00434843
Lead, foil, 300x300mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF87346806
Lead, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF50662773
Lead, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF98077898
Lead, foil, 1000x1000mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF68213538
Lead, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF32778322
Lead, foil, 100x100mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF21778397
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF54864041
Lead, foil, 100x100mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF64074034
Lead, foil, 300x300mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF06345479
Lead, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Pb