EMAIL THIS PAGE TO A FRIEND
Amino acids
Asymmetric synthesis of all stereoisomers of 6-methylpipecolic acids.
PMID 12707808
Abstract
Asymmetric synthesis of all four stereoisomers of 6-methylpipecolic acids with high enantiomeric purity via iterative AD reaction, starting from 1,6-heptadiene, has been described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
258849
1,6-Heptadiene, 99%
C7H12