EMAIL THIS PAGE TO A FRIEND
The Journal of steroid biochemistry and molecular biology
Glucocorticoid metabolism by 11-beta hydroxysteroid dehydrogenase type 2 modulates human mineralocorticoid receptor transactivation activity.
PMID 12711009
Abstract
The mineralocorticoid receptor (MR) binds aldosterone, but also glucocorticoid hormones (corticosterone in rodents, cortisol in humans), which largely prevail in the plasma. To prevent permanent and maximal occupancy of MR by glucocorticoid hormones in aldosterone-target cells, specific effects of aldosterone require metabolism of glucocorticoid hormones into 11-dehydroderivatives by 11-beta hydroxysteroid dehydrogenase (11-HSD2). We analyzed the effect of corticosterone or 11-dehydrocorticosterone (11-DHC) on the transactivation activity of the MR, transiently expressed in a new renal cell line expressing 11-HSD2. We show that, because of its metabolism by 11-HSD2, corticosterone is a poor activator of MR transactivation, except at micromolar concentrations, where the enzyme is saturated. We also show that high micromolar concentrations of 11 DHC are required to activate the MR. The weak antagonist property of 11-DHC on aldosterone-induced hMR transactivations is also documented. Such partial agonist activity of 11-DHC is discussed in the light of its positioning in a three-dimensional model of the MR ligand-binding domain.