EMAIL THIS PAGE TO A FRIEND
Organic letters
Fluorescence sensing of ammonium and organoammonium ions with tripodal oxazoline receptors.
PMID 12713288
Abstract
A new class of fluorescence sensors for ammonium and organoammonium ions has been disclosed. One of the sensors, an alaninol-derived tripodal oxazoline (1a) shows significant fluorescence enhancement upon binding NH(4)(+) but little response toward K(+), Na(+), and Mg(2+) ions. Owing to its chiral environment, a phenylglycinol-derived tripodal oxazoline (1b) shows chiral discrimination in fluorescence upon binding enantiomeric guests. [reaction: see text]