EMAIL THIS PAGE TO A FRIEND
Canadian journal of microbiology
Melanin biosynthesis in the fungus Curvularia lunata (teleomorph: Cochliobolus lunatus).
PMID 12718399
Abstract
Curvularia lunata (teleomorph: Cochliobolus lunatus) is a known plant and human pathogen. Tricyclazole, a specific inhibitor of pentaketide melanin biosynthesis, blocked the biosynthesis of melanin in Curvularia lunata and caused the accumulation of the melanin metabolites flaviolin and 2-hydroxyjuglone. This showed that melanin in Curvularia lunata is produced by a pentaketide pathway from 1,8-dihydroxynaphthalene. The 1,3,8-trihydroxynaphthalene reductase (3HNR) gene, associated with the melanin pathway of Curvularia lunata, was identified and characterized. An alignment of 3HNR sequences enabled the design of primers covering conserved regions. A PCR-amplified fragment of Curvularia lunata genomic DNA was used for screening the cDNA library. Three independent cDNA clones revealed an 801-bp open reading frame encoding a 267 amino acid protein. The protein was expressed in Escherichia coli and purified to homogeneity. The predicted amino acid sequence of the 28.6-kDa protein demonstrated homology to other fungal 3HNR and other members of the short-chain dehydrogenase super family. Northern analyses revealed that 3HNR from Curvularia lunata is expressed synchronously with melanization after 3 days of Curvularia lunata growth in malt extract medium. No 3HNR reductase gene expression nor melanization was observed when Curvularia lunata was grown in yeast nitrogen base medium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45808
Tricyclazole, PESTANAL®, analytical standard
C9H7N3S