EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Adeno-associated viral transfer of opioid receptor gene to primary sensory neurons: a strategy to increase opioid antinociception.
PMID 12719538