EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Identification of the L-aspartate transporter in Bacillus subtilis.
PMID 12730183
Abstract
YveA of Bacillus subtilis, a putative transporter of the amino acid/polyamine/organocation (APC) superfamily, is shown to mediate uptake of both L-aspartate and L-glutamate as well as having sensitivity to L-aspartate hydroxamate. This 14 TMS protein is the primary aspartate uptake system in B. subtilis and serves as the prototype for a new family within the APC superfamily.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6508
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
C4H8N2O4