EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of carbohydrates as their 3-aminophthalhydrazide derivatives by capillary zone electrophoresis with on-line chemiluminescence detection.
PMID 12735474
Abstract
A method based on pre-capillary derivatization with luminol (3-aminophthalhydrazide) for carbohydrate analysis using capillary electrophoresis with on-line chemiluminescence (CL) detection was developed. The derivatives of seven monosaccharides were separated and detected by using 200 mM borate buffer containing 100 mM hydrogen peroxide at pH 10.0 as separation electrolyte and 25 mM hexacyanoferrate in 3 M sodium hydroxide solution as post-capillary chemiluminescence reagent with separation efficiencies ranging from 160,000 to 231,000 plates per metre. The minimum amount of carbohydrate derivatized was 2 pmol (corresponding to the concentration of 2 microM). The method also provided a linear response for glucose in the concentration range of 0.1-250 microM with a mass detection limit of 420 amol or a concentration detection limit of 0.1 microM. Preliminary work using the CE-CL format to determine glucose in a rat brain microdialysis sample is presented as a typical case.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8264
4-Aminophthalhydrazide
C8H7N3O2