EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Palladium-catalysed dimerization of vinylarenes using indium triflate as an effective co-catalyst.
PMID 12739644
Abstract
A palladium-indium triflate catalyst was found to be much more active for the dimerization of vinylarenes compared with generally used cationic palladium(II) catalysts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442151
Indium(III) trifluoromethanesulfonate
C3F9InO9S3