EMAIL THIS PAGE TO A FRIEND
Molecular membrane biology
Influence of erythrocyte shape on the rate of Ca2+-induced scrambling of phosphatidylserine.
PMID 12745928
Abstract
Membrane-perturbing agents that cause transformation of biconcave erythrocytes into echinocytes or stomatocytes were used to investigate the influence of erythrocyte shape on the rate of Ca(2+)-induced scrambling of phospholipids. Erythrocytes were treated with a variety of lipid-soluble compounds to induce these shape changes, followed by incubation with calcium and ionomycin to activate lipid scramblase. Prothrombinase activity of the cells was used to monitor the rate of surface exposure of phosphatidylserine, which is taken as a measure of scramblase activity. Echinocytes show an enhanced rate of scrambling, whereas stomatocytes show a reduced rate, relative to normocytes. This phenomenon appears to correlate with enhanced and diminished micro-exovesicle shedding from echinocytes and stomatocytes, respectively. It is concluded that the rate of calcium-induced phosphatidylserine exposure (rate of lipid scrambling) in erythrocytes depends for a considerable part on the cells' ability to form microvesicles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7081
1,2-Didecanoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)
C28H56NO8P