EMAIL THIS PAGE TO A FRIEND
Mycological research
Amanita muscaria: chemistry, biology, toxicology, and ethnomycology.
PMID 12747324
Abstract
The fly agaric is a remarkable mushroom in many respects; these are its bearing, history, chemical components and the poisoning that it provokes when consumed. The 'pantherina' poisoning syndrome is characterized by central nervous system dysfunction. The main species responsible are Amanita muscaria and A. pantherina (Amanitaceae); however, some other species of the genus have been suspected for similar actions. Ibotenic acid and muscimol are the active components, and probably, some other substances detected in the latter species participate in the psychotropic effects. The use of the mushroom started in ancient times and is connected with mysticism. Current knowledge on the chemistry, toxicology, and biology relating to this mushroom is reviewed, together with distinctive features concerning this unique species.