EMAIL THIS PAGE TO A FRIEND
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan
Effects of exogenous polyamines on growth, toxicity, and toxin profile of dinoflagellate Alexandrium minutum.
PMID 12749197
Abstract
The effect of exogenous polyamines (cadaverine, putrescine, norspermidine, spermidine, and spermine) on the growth, toxicity, and toxin profile of the dinoflagellate Alexandrium minutum T1 was examined. It was found that cadaverine at concentrations of 0.1-2.0 mumol/L enhanced the growth of A. minutum T1. Putrescine and norspermidine at a low level (0.1 mumol/L) also promoted the algal growth. Spermidine depressed the algal growth. However, the cell toxicity levels of A. minutum T1 cultured with or without cadaverine, putrescine, norspermidine, and spermidine were almost the same. The toxic components of A. minutum T1 were GTXs 1-4 only, and GTXs 1 and 4 were predominant (74.6 +/- 7.1%) in all cultures. On the other hand, spermine did not effect the growth of A. minutum T1, though it decreased the cell toxicity and the ratio of GTX 2 + GTX 3 (15.0 +/- 6.6%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14520
Bis(3-aminopropyl)amine, purum, ≥97.0% (GC)
C6H17N3
I1006
Bis(3-aminopropyl)amine, 98%
C6H17N3