EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Redefining the structure-activity relationships of 2,6-methano-3-benzazocines. Part 2: 8-formamidocyclazocine analogues.
PMID 12749896
Abstract
High affinity binding for mu and kappa opioid receptors has been observed in analogues of cyclazocine, ethylketocyclazocine and naltrexone where the prototypic (of opiates) phenolic OH group was replaced with a formamide (-NHCHO) group. For the 8-formamide analogue of cyclazocine, binding is highly enantiospecific (eudismic ratios approximately 2000 for mu and kappa) with K(i) values