EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis due to DL-alpha-tocopheryl nicotinate.
PMID 12755738
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5134
(±)-α-Tocopherol nicotinate
C35H53NO3