EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Structure-toxicity relationships for the effects of N- and N,N'-alkyl thioureas to Tetrahymena pyriformis.
PMID 12756468
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M84607
N-Methylthiourea, 97%
C2H6N2S