EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Spectrophotometric assay of yeast vitality using 2,3,5,6-tetramethyl-1,4-benzoquinone and tetrazolium salts.
PMID 12769360
Abstract
A method for the spectrophotometric assay of yeast vitality was developed using 2,3,5,6-tetramethyl-1,4-benzoquinone and tetrazolium salts. The metabolic efficiency of 2,3,5,6-tetramethyl-1,4-benzoquinone by yeast cells was used as an index of yeast vitality. 2,3,5,6-Tetramethyl-1,4-benzoquinone was reduced to 2,3,5,6-tetramethyl-1,4-hydroquinone by yeast cells. Then, the superoxide anion radicals generated from O2 by reduction with 2,3,5,6-tetramethyl-1,4-hydroquinone under alkaline conditions reduced tetrazolium salts to formazan, which exhibited absorbance maxima at 440 nm. A linear relationship between the absorbance and viable cell density was obtained in the range of 1.0 x 10(5)-2.0 x 10(7) cells/ml for a sample solution. During the cultivation of yeast cells, the absorbance showed almost an anti-parallel change with that of glucose in yeast growth and fermentation, suggesting that the absorbance change reflected the vitality of yeast cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06856
Duroquinone, Standard for quantitative NMR, TraceCERT®
C10H12O2
D223204
Duroquinone, 97%
C10H12O2