EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Models of acute inflammation in the ear.
PMID 12769482
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E45309
Ethyl phenylpropiolate, 98%
C11H10O2