EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on chemical constituents of two plants from Costus].
PMID 12774385
Abstract
To study the chemical constituents of Costus speciosus and C. tonkinensis (Zingiberaceae) distributed in Yunnan province. Chromatography and spectral analyses were used to isolate the constituents and elucidate their structure. Six compounds were isolated from the rhizome of C. speciosus and elucidated as diosgenin(1), prosapogenin B of dioscin(2), diosgenone(3), cycloartanol(4), 25-en-cycloartenol(5) and octacosanoic acid(6). Four compounds were isolated from the rhizome of Costus tonkinensis and elucidated as tetracosanoic acid(7), succinic acid(8), beta-sitosterol(9) and daucosterin(10). Compounds of 3-6 were obtained from C. speciosus for the first time and compounds of 7-10 were obtained from C. tonkinensis for the first time too.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284432
Octacosanoic acid, synthetic, ≥98%
C28H56O2