EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on chemical constituents of Umbilicaria esculenta (Miyoshi) minks].
PMID 12776427
Abstract
To study the chemical constituents from the fruit bodies of Umbilicaria esculenta. Five compounds were isolated from the ethanolic extract of this plant by all kinds of column chormatography, and structures were identified by spectroscopic analysis. Five compounds were identified as ethyl orsellinate, orsellinic acid, orcinol, lecanoric acid and lecanorin. Ethyl orsellinate, orsellinic acid, orcinol and lecanorin were isolated from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
447420
Orcinol, 97%
C7H8O2