EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Field test and lab experiment on control efficacy of the pathogen of opium poppy mildew].
PMID 12776513
Abstract
To screen effectual fungicides for field control because of the seriousness of opium poppy mildew and importance of chemical control on plant diseases. Seven fungicides were screened in Lab experiment and field test during 1996-1997. All of them and their different dosages were effective to control conidia of Peronospora arborescens. Among them, 72.2% propamocarb of 1203 and 902.5 ppm were the most effective both in Lab experiment and field test with efficacy 79.91% and 79.33% respectively in field test, and the efficacy of other fungicides was over 50%. Seven fungicides tested can be used to control nonsystematic symptom of opium poppy mildew.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45638
Propamocarb, PESTANAL®, analytical standard
C9H20N2O2