EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[A study on chemical constituents in the herb of Mentha spicata].
PMID 12776552
Abstract
To investigate the constituents of anti-inflammatory and hemostatic active sites of Mentha spicata. Chemical constituents were separated by solvent method and chromatography and identified by physicochemical properties and spectroscopic methods. The structures were identified as ursane I, 3-methoxy-4-methylbenzaldehyde II, veratric acid III, 5-hydroxy-3',4',6,7-tetramethoxyflavone IV, diosmetin(V), thymonin VI, daucosterol VII. Among them, compounds I, II, III were first obtained from M. genus, IV, VI were separated from M. spicta for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D131806
3,4-Dimethoxybenzoic acid, ≥99%
C9H10O4