EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A novel bioassay for screening and quantification of taxanes.
PMID 12778728
Abstract
We report a novel method for screening and quantifying tubulin-binding antimitotic agents that promote microtubule formation. The method is based on the shift in the peak of the fluorescence emission difference spectrum of tubulin complexed to metal free tetrakis(4-carboxyphenyl)porphyrin (TCPP) in the presence of antimitotic agents. Taxol (paclitaxel), an anti-tumor drug approved for the treatment of a variety of cancers, caused the appearance of a new fluorescence peak at 645 nm at concentrations as low as 125 nM, the intensity of which was a function of the paclitaxel concentration. Cephalomannine caused the induction of a new fluorescence peak at 651 nm only above 1 muM. Baccatin did not induce the appearance of any new peak within detectable operating measurement conditions. These observations are in accordance with the biological activities/cytotoxicities of these compounds. Accordingly, it is proposed that the new method can be used for high throughput screening of antimitotic compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4991
Cephalomannine, ≥97% (HPLC), solid
C45H53NO14