EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
Continuous cultivations of a Penicillium chrysogenum strain expressing the expandase gene from Streptomyces clavuligerus: Growth yields and morphological characterization.
PMID 12783492
Abstract
The growth stoichiometry of a Penicillium chrysogenum strain expressing the expandase gene from Streptomyces clavuligerus was determined in glucose-limited chemostat cultivations using a chemically defined medium. This strain produces adipoyl-7-aminodeacetoxycephalosporanic acid (ad-7-ADCA) when it is fed with adipic acid. The biomass yield and maintenance coefficients for the strain were similar to those found for penicillin-producing strains of Penicillium chrysogenum. The maximum specific growth rate in the chemostat was found to be 0.11 h(-1). Metabolic degradation of adipate was found to take place in significant amounts only at dilution rates below 0.03 h(-1). After three to five residence times, adipate degradation and ad-7-ADCA production disappeared, and this allowed determination of the biomass yield coefficient on adipate. The morphology was measured at different dilution rates and the mean total hyphal length and mean number of tips both increased with an increase in dilution rate from 0.015 to 0.065 h(-1). Both variables decreased when the dilution rate was increased above 0.065 h(-1). A correlation between mean total hyphal length and productivity of ad-7-ADCA was found.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8398
7-Aminodesacetoxycephalosporanic acid
C8H10N2O3S