EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Catalytic alcoholysis of tetramethylsilane via Pt-mediated C-H bond activation.
PMID 12785766
Abstract
Tetramethylsilane reacts with 2,2,2-trifluoroethanol (TFE) in the presence of a cationic platinum(II) catalyst [(NN)PtMe(TFE)]+ (NN = 1,2-bis(3,5-dimethylphenylimino)butane). Catalytic Si-C bond heterolysis results in the formation of the trimethylsilyl ether, Me3SiOCH2CF3, accompanied by liberation of one equivalent of methane. Preliminary experiments suggest that a rate-determining C-H bond activation precedes rapid attack by solvent at silicon to yield the silyl ether product and regenerate the active platinum methyl cation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T24007
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
C4H12Si