EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Patch testing with zinc dibenzyldithiocarbamate. A multicentre study of the Information Network of Departments of Dermatology and the German Contact Dermatitis Research Group.
PMID 12786726
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329711
Zinc dimethyldithiocarbamate, 97%
C6H12N2S4Zn
45708
Ziram, PESTANAL®, analytical standard
C6H12N2S4Zn