EMAIL THIS PAGE TO A FRIEND
Mutation research
The unusual effect of pKM101 on the mutagenicity of acetaldehyde oxime in Salmonella typhimurium.
PMID 12787823
Abstract
Acetaldehyde oxime was found to induce more revertants in Salmonella typhimurium strain TA1535 than in TA100 in the absence of S9 metabolic activation. TA100 was originally constructed from TA1535 by the addition of the plasmid pKM101, carrying mucAB which generally enhances sensitivity to the mutagenic effects of chemicals. The role of pKM101 in lowering the sensitivity to acetaldehyde oxime was explored by: (1) increasing the incubation time of the selective agar plates from 2 to 3 days; (2) using a new strain, isogenic to TA100, constructed by introducing pKM101 into the TA1535 isolate used in these experiments; (3) by testing a strain constructed by inserting into TA1535 a plasmid carrying mucAB but otherwise unrelated to pKM101. Each of these alterations increased the number of revertants per plate in the presence of acetaldehyde oxime, indicating that the apparent nonmutagenicity of this chemical in TA100 is due to multiple factors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1002
Acetaldehyde oxime, mixture of syn and anti, 99%
C2H5NO