EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis, isolation, and characterization of 2'-paclitaxel glycinate: an application of the Bsmoc protecting group.
PMID 12790596
Abstract
The relevance of the Bsmoc protecting group in the synthesis of moisture- and base-sensitive compounds has been demonstrated by the preparation of 2'-paclitaxel glycinate with use of the solid anhydrous base, 4-piperidinopiperidine in DCM solvent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
534498
4-Piperidinopiperidine, 97%
C10H20N2