EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Pharmaceutical profiling method for lipophilicity and integrity using liquid chromatography-mass spectrometry.
PMID 12798198
Abstract
A method is described for the simultaneous profiling of sample lipophilicity, integrity, and purity. The method is rapid and is applicable to high throughput profiling of pharmaceutical properties in drug discovery. A short Polaris C(18) column is used with a rapid, wide-polarity mobile phase gradient, UV detection, and MS analysis. The lipophilicity of each component is estimated from a calibration curve using six drug or organic compounds and plotting their respective measured retention time versus LogD(7.4) (literature). The correlation of LogD(7.4) (literature) to LogD(7.4) (HPLC) for 60 structurally diverse drugs has a correlation coefficient r(2) of 0.89. The method is applicable to compounds with MW>200 and retention time>1.5 min for rapid, initial pharmaceutical profiling in drug discovery.