EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Effects of dynorphin A(1-13) on DNA synthesis and free radical oxidation in respiratory organs of newborn albino rats pretreated with NG-nitro-L-arginine methyl ester.
PMID 12802389
Abstract
Dynorphin A(1-13) increased the number of DNA-synthesizing epitheliocytes in the bronchi and inhibited free radical oxidation in the lungs of newborn albino rats. Pretreatment with NG-nitro-L-arginine methyl ester not only abolished, but also reversed the effect of this peptide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7017
Dynorphin A Porcine Fragment 1-13, ≥97% (HPLC)
C75H126N24O15