EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on linalyl acetate.
PMID 12804651
Abstract
A toxicologic and dermatologic review of linalyl acetate when used as a fragrance ingredient is presented.