EMAIL THIS PAGE TO A FRIEND
FEBS letters
Motilin-bicelle interactions: membrane position and translational diffusion.
PMID 12804764
Abstract
The interaction between the peptide hormone motilin and bicelles has been investigated by pulsed field gradient-nuclear magnetic resonance methods and by the use of paramagnetic probes. Diffusion coefficients were measured for motilin, the phospholipids with and without motilin, and for tetramethylsilane. The results show that around 90% of motilin is bound to acidic bicelles and 84% of motilin is bound to neutral bicelles. It is found that the apparent bicelle size is reduced by the presence of motilin. This cannot be explained by changes in 1,2-dihexanoyl-sn-glycero-3-phosphatidylcholine solubility. The use of paramagnetic agents to investigate the position of motilin shows that the turn in the N-terminus of motilin is inserted into the bicelle, while the helix most likely resides within the head-group layer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T24007
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
C4H12Si