EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Acetohydroxyacid synthase from Mycobacterium avium and its inhibition by sulfonylureas and imidazolinones.
PMID 12818195
Abstract
Tuberculosis (TB) remains one of the world's leading causes of death from infectious disease. It is caused by infection with Mycobacterium tuberculosis or sometimes, particularly in immune-compromised patients, Mycobacterium avium. The aim of this study was to create a tool that could be used in the search for new anti-TB drugs that inhibit branched-chain amino acid (BCAA) biosynthesis, as these are essential amino acids that are not available to a mycobacterium during growth in an infected organism. To this end, we cloned, overexpressed, purified and characterised for the first time an acetohydroxyacid synthase (AHAS), a key enzyme in the pathway to the biosynthesis of the BCAAs, from the genus Mycobacterium. Nine commercial herbicides of the sulfonylurea and imidazolinone classes were tested for their influence on this enzyme. Four of the sulfonylureas were potent inhibitors of the enzyme. The relative potency of the different inhibitors towards the M. avium enzyme was unlike their potency towards other AHASs whose inhibitor profile has been reported, emphasising the advantage of using a mycobacterial enzyme as a tool in the search for new anti-TB drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34322
Chlorosulfuron, PESTANAL®, analytical standard
C12H12ClN5O4S
33383
Triasulfuron, PESTANAL®, analytical standard
C14H16ClN5O5S