EMAIL THIS PAGE TO A FRIEND
Nederlands tijdschrift voor geneeskunde
[Methemoglobinemia due to ingestion of isobutyl nitrite ('poppers')].
PMID 12822531
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
327190
Isobutyl nitrite, 95%
C4H9NO2